missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzaldehyde, 98%

Catalog No. AAL1286609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1286609 10 g
AAL1286618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1286609 Supplier Thermo Scientific Chemicals Supplier No. L1286609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5896-17-3 | C14H12O2 | 212.25 g/mol

2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde
PubChem CID 344784
IUPAC Name 2-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 46°C to 47°C
Quantity 10 g
Beilstein 780307
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 2-Benzyloxybenzaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only