missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylimidazoline 97.0+%, TCI America™

Catalog No. B38065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38065G Supplier TCI America Supplier No. B38065G
Only null left

Chemical Identifiers

CAS 59-98-3
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005182
InChI Key JIVZKJJQOZQXQB-UHFFFAOYSA-N
Synonym Tolazoline
PubChem CID 5504
ChEBI CHEBI:28502
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole
SMILES C1CN=C(N1)CC2=CC=CC=C2

Specifications

Melting Point 66°C
Boiling Point 188°C
Quantity 5 g
Formula Weight 160.22
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Benzylimidazoline
EINECSNumber (9)-1346
RTECSNumber NJ1925000
TSCA No
Recommended Storage Refrigerator