missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylbenzimidazole Hydrochloride 98.0+%, TCI America™

Catalog No. p-7123481
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43415G 5 g
B434125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43415G Supplier TCI America Supplier No. B43415G
Only null left

Chemical Identifiers

CAS 1212-48-2
Molecular Formula C14H13ClN2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00443659
InChI Key CJTQARUHALKPGG-UHFFFAOYSA-N
PubChem CID 164798
IUPAC Name hydrogen 2-benzyl-1H-1,3-benzodiazole chloride
SMILES [H+].[Cl-].C(C1=NC2=CC=CC=C2N1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 244.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylbenzimidazole Hydrochloride