missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzylaminopyridine 98.0+%, TCI America™
$80.16 - $594.63
Chemical Identifiers
| CAS | 6935-27-9 |
|---|---|
| Molecular Formula | C12H12N2 |
| Molecular Weight (g/mol) | 184.242 |
| MDL Number | MFCD00006251 |
| InChI Key | WYHXNQXDQQMTQI-UHFFFAOYSA-N |
| Synonym | 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g |
| PubChem CID | 23362 |
| IUPAC Name | N-benzylpyridin-2-amine |
| SMILES | C1=CC=C(C=C1)CNC2=CC=CC=N2 |
Chemical Identifiers
| 6935-27-9 | |
| 184.242 | |
| WYHXNQXDQQMTQI-UHFFFAOYSA-N | |
| 23362 | |
| C1=CC=C(C=C1)CNC2=CC=CC=N2 |
| C12H12N2 | |
| MFCD00006251 | |
| 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g | |
| N-benzylpyridin-2-amine |
Specifications
| 6935-27-9 | |
| 182°C | |
| MFCD00006251 | |
| 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g | |
| C1=CC=C(C=C1)CNC2=CC=CC=N2 | |
| 184.242 | |
| 184.24 | |
| Crystalline Powder |
| 95°C | |
| C12H12N2 | |
| 25 g | |
| WYHXNQXDQQMTQI-UHFFFAOYSA-N | |
| N-benzylpyridin-2-amine | |
| 23362 | |
| ≥98.0% (GC,T) | |
| 2-Benzylaminopyridine |
Safety and Handling
TSCA : Yes