missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylacrylic Acid 98.0+%, TCI America™

Catalog No. B34095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34095G 5 g
B340925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34095G Supplier TCI America Supplier No. B34095G
Only null left

Chemical Identifiers

CAS 5669-19-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00854174
InChI Key RYNDYESLUKWOEE-UHFFFAOYSA-N
Synonym 2-Methylene-3-phenylpropionic Acid
PubChem CID 303571
IUPAC Name 2-benzylprop-2-enoic acid
SMILES OC(=O)C(=C)CC1=CC=CC=C1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylacrylic Acid