missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. p-7122953
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34481G 1 g
B34485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34481G Supplier TCI America Supplier No. B34481G
Only null left

Chemical Identifiers

CAS 87100-28-5
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.103
MDL Number MFCD05663841
InChI Key YCNQPAVKQPLZRS-UHFFFAOYSA-N
Synonym benzylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl, acmc-209qek, benzylboronic pinacol ester, benzyl boronic acid pinacol ester, benzylboronic acid, pinacol ester, 2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane
PubChem CID 3864964
IUPAC Name 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 g
Formula Weight 218.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane