missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylthiophene 98.0+%, TCI America™

Catalog No. B030825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B030825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B030825G Supplier TCI America Supplier No. B030825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-00-2
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD00022495
InChI Key DWYFUJJWTRPARQ-UHFFFAOYSA-N
Synonym 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
PubChem CID 67262
IUPAC Name phenyl(thiophen-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 56°C
Boiling Point 300°C
Quantity 25 g
Formula Weight 188.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylthiophene