missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylthiophene, 98%

Catalog No. AAA1203618
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1203618 50 g
AAA1203606 5 g
AAA1203609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1203618 Supplier Thermo Scientific Chemicals Supplier No. A1203618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135-00-2 | C11H8OS | 188.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-00-2
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD00022495
InChI Key DWYFUJJWTRPARQ-UHFFFAOYSA-N
Synonym 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
PubChem CID 67262
IUPAC Name phenyl(thiophen-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 55°C to 57°C
Boiling Point 124°C to 126°C (0.5 mmHg)
Quantity 50 g
Beilstein 3929
Formula Weight 188.25
Percent Purity 98%
Chemical Name or Material 2-Benzoylthiophene
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 205-169-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only