missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzoylpyridine 99.0+%, TCI America™
$33.56 - $430.60
Chemical Identifiers
| CAS | 91-02-1 |
|---|---|
| Molecular Formula | C12H9NO |
| Molecular Weight (g/mol) | 183.21 |
| MDL Number | MFCD00006300 |
| InChI Key | GCSHUYKULREZSJ-UHFFFAOYSA-N |
| Synonym | 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone |
| PubChem CID | 7038 |
| IUPAC Name | phenyl(pyridin-2-yl)methanone |
| SMILES | C1=CC=C(C=C1)C(=O)C2=CC=CC=N2 |
Chemical Identifiers
| 91-02-1 | |
| 183.21 | |
| GCSHUYKULREZSJ-UHFFFAOYSA-N | |
| 7038 | |
| C1=CC=C(C=C1)C(=O)C2=CC=CC=N2 |
| C12H9NO | |
| MFCD00006300 | |
| 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone | |
| phenyl(pyridin-2-yl)methanone |
Specifications
| 91-02-1 | |
| White-Yellow | |
| MFCD00006300 | |
| 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone | |
| C1=CC=C(C=C1)C(=O)C2=CC=CC=N2 | |
| 183.21 | |
| 183.21 | |
| Crystalline Powder |
| 44°C | |
| C12H9NO | |
| 25 g | |
| GCSHUYKULREZSJ-UHFFFAOYSA-N | |
| phenyl(pyridin-2-yl)methanone | |
| 7038 | |
| ≥99.0% (GC,T) | |
| 2-Benzoylpyridine |
Safety and Handling
RTECSNumber : OB6400000
TSCA : Yes