missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylpyridine, 99+%

Catalog No. AC105800250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105800250 25 g Glass bottle
AC105801000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105800250 Supplier Thermo Scientific Chemicals Supplier No. 105800250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-02-1 | C12H9NO | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-02-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006300
InChI Key GCSHUYKULREZSJ-UHFFFAOYSA-N
Synonym 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
PubChem CID 7038
IUPAC Name phenyl(pyridin-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=N2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 42.0°C to 44.0°C
Color Colorless to Yellow
Boiling Point 317.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5CO(C5H4N)
Quantity 25 g
Beilstein 21, 330
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 183.21
Percent Purity 99+%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Benzoylpyridine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 202-034-3
RTECSNumber OB6400000
TSCA TSCA

RUO – Research Use Only