missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylbenzoic Acid, 98+%, Thermo Scientific™

Catalog No. p-63643
Change view
Click to view available options
Quantity:
25g
500g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105710250 25g Plastic bottle
AC105715000 500g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105710250 Supplier Thermo Scientific Chemicals Supplier No. 105710250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 85-52-9
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002472
InChI Key FGTYTUFKXYPTML-UHFFFAOYSA-N
Synonym o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
PubChem CID 6813
IUPAC Name 2-benzoylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Boiling Point 257.0°C to 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula C6H5COC6H4CO2H
Quantity 25g
Beilstein 10, 747
Formula Weight 226.23
Percent Purity 98+%
Chemical Name or Material 2-Benzoylbenzoic acid, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-612-2
RTECSNumber DG3600000
TSCA TSCA

RUO – Research Use Only