missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, 98%

Catalog No. AC169085000
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169085000 500 g Glass bottle
AC169080250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169085000 Supplier Thermo Scientific Chemicals Supplier No. 169085000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-49-4 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-49-4
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00005716
InChI Key ASSKVPFEZFQQNQ-UHFFFAOYSA-N
Synonym 2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
PubChem CID 6043
IUPAC Name 3H-1,3-benzoxazol-2-one
SMILES O=C1NC2=CC=CC=C2O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 137°C to 141°C
Color Beige to Brown-Gray
Density 160g/mL
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in ethanol,ethyl ether and,trifluoroacetic acid
Formula Weight 135.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Benzoxazolinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-430-

RUO – Research Use Only