missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, 98%

Catalog No. AAA1937814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1937814 25 g
AAA1937822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1937814 Supplier Thermo Scientific Chemicals Supplier No. A1937814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59-49-4 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-49-4
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00005716
InChI Key ASSKVPFEZFQQNQ-UHFFFAOYSA-N
Synonym 2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
PubChem CID 6043
IUPAC Name 2,3-dihydro-1,3-benzoxazol-2-one
SMILES O=C1NC2=CC=CC=C2O1

Specifications

Melting Point 139°C to 140°C
Flash Point 160°C (320°F)
Quantity 25 g
Beilstein 119481
Formula Weight 135.12
Percent Purity 98%
Chemical Name or Material 2-Benzoxazolinone
missing translation for 'healthHazard1' H302+H312+H332
missing translation for 'healthHazard2' GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 200-430-0
RTECSNumber DM4905000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only