missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Benzimidazolyl)acetonitrile 96.0+%, TCI America™
$82.02 - $281.48
Chemical Identifiers
| CAS | 4414-88-4 |
|---|---|
| Molecular Formula | C9H7N3 |
| Molecular Weight (g/mol) | 157.176 |
| InChI Key | BWOVACANEIVHST-UHFFFAOYSA-N |
| Synonym | 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile |
| PubChem CID | 20455 |
| IUPAC Name | 2-(1H-benzimidazol-2-yl)acetonitrile |
| SMILES | C1=CC=C2C(=C1)NC(=N2)CC#N |
Chemical Identifiers
| 4414-88-4 | |
| 157.176 | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| 2-(1H-benzimidazol-2-yl)acetonitrile |
| C9H7N3 | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 20455 | |
| C1=CC=C2C(=C1)NC(=N2)CC#N |
Specifications
| 4414-88-4 | |
| C9H7N3 | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| C1=CC=C2C(=C1)NC(=N2)CC#N | |
| 157.176 | |
| 157.18 | |
| Crystalline Powder |
| 212°C | |
| 5 g | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 2-(1H-benzimidazol-2-yl)acetonitrile | |
| 20455 | |
| ≥96.0% (GC,T) | |
| (2-Benzimidazolyl)acetonitrile |
Safety and Handling
RTECSNumber : DD5650000
TSCA : Yes