missing translation for 'onlineSavingsMsg'
Learn More

2-Benzimidazoleacetonitrile, 99%

Catalog No. AAL0082718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0082718 50 g
AAL0082709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0082718 Supplier Thermo Scientific Chemicals Supplier No. L0082718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4414-88-4 | C9H7N3 | 157.176 g/mol

2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 4414-88-4
Molecular Formula C9H7N3
Molecular Weight (g/mol) 157.176
MDL Number MFCD00005601
InChI Key BWOVACANEIVHST-UHFFFAOYSA-N
Synonym 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile
PubChem CID 20455
IUPAC Name 2-(1H-benzimidazol-2-yl)acetonitrile
SMILES C1=CC=C2C(=C1)NC(=N2)CC#N

Specifications

Melting Point 206°C to 211°C
Odor Odorless
Quantity 50 g
Beilstein 128461
Solubility Information Insoluble in water.
Formula Weight 157.18
Percent Purity 99%
Chemical Name or Material 2-Benzimidazoleacetonitrile
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 224-574-9
RTECSNumber DD5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only