missing translation for 'onlineSavingsMsg'
Learn More

2-Azidoethyl 2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranoside 92.0+%, TCI America™

Catalog No. p-7117871
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23771G 1 g
A23775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23771G Supplier TCI America Supplier No. A23771G
Only null left

Chemical Identifiers

CAS 140428-81-5
Molecular Formula C16H23N3O10
Molecular Weight (g/mol) 417.371
InChI Key DPAXDKFCBLGMIZ-WFDRTZIDSA-N
PubChem CID 53384426
IUPAC Name [(3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(2-azidoethoxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OCCN=[N+]=[N-])OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 115°C
Color White
Quantity 1 g
Formula Weight 417.37
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Azidoethyl 2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranoside
TSCA No
Recommended Storage Refrigerator