missing translation for 'onlineSavingsMsg'
Learn More

2'-Azido-2'-deoxyuridine 98.0+%, TCI America™

Catalog No. A294225MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A294225MG 25 mg
A2942100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A294225MG Supplier TCI America Supplier No. A294225MG

Chemical Identifiers

CAS 26929-65-7
Molecular Formula C9H11N5O5
Molecular Weight (g/mol) 269.217
MDL Number MFCD00043045
InChI Key MRUKYOQQKHNMFI-XVFCMESISA-N
Synonym 2'-azido-2'-deoxyuridine, n3-du, uridine,2'-azido-2'-deoxy, 2 inverted exclamation marka-azido-2 inverted exclamation marka-deoxyuridine, 1-2r,3r,4s,5r-3-azido-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-azido,2'-deoxyuridine, 2'-azido-2'-desoxyuridine, 2'-deoxy-2'-azido-uridine, 2'-azido-2'-deoxy uridine, uridine, 2'-azido-2'-deoxy
PubChem CID 168629
IUPAC Name 1-[(2R,3R,4S,5R)-3-azido-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)N=[N+]=[N-]

Specifications

Melting Point 151°C
Color White
Quantity 25 mg
Formula Weight 269.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Azido-2′-deoxyuridine
TSCA No
Recommended Storage Refrigerator