missing translation for 'onlineSavingsMsg'
Learn More

2-Azabicyclo[2.2.1]hept-5-en-3-one 98.0+%, TCI America™

Catalog No. p-7117388
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13591G 1 g
A13595G 5 g
A135925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A13591G Supplier TCI America Supplier No. A13591G

Chemical Identifiers

CAS 49805-30-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00213364
InChI Key DDUFYKNOXPZZIW-UHFFFAOYSA-N
Synonym 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one
PubChem CID 548678
IUPAC Name 3-azabicyclo[2.2.1]hept-5-en-2-one
SMILES C1C2C=CC1NC2=O

Specifications

Melting Point 57°C
Quantity 1 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Azabicyclo[2.2.1]hept-5-en-3-one
EINECSNumber (5)-6630
TSCA No
Recommended Storage Refrigerator