Learn More
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific™
$58.41 - $363.31
Chemical Identifiers
| CAS | 49805-30-3 |
|---|---|
| Molecular Formula | C6H7NO |
| Molecular Weight (g/mol) | 109.13 |
| MDL Number | MFCD00213364 |
| InChI Key | DDUFYKNOXPZZIW-UHFFFAOYSA-N |
| Synonym | 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one |
| PubChem CID | 548678 |
| IUPAC Name | 3-azabicyclo[2.2.1]hept-5-en-2-one |
| SMILES | C1C2C=CC1NC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC297060010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC297060100
|
N/A | 10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 49805-30-3 | |
| 109.13 | |
| DDUFYKNOXPZZIW-UHFFFAOYSA-N | |
| 548678 | |
| C1C2C=CC1NC2=O |
| C6H7NO | |
| MFCD00213364 | |
| 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one | |
| 3-azabicyclo[2.2.1]hept-5-en-2-one |
Specifications
| 49805-30-3 | |
| 97.5 | |
| 102°C to 106°C (0.2 hPa) | |
| Authentic | |
| Glass bottle | |
| MFCD00213364 | |
| Solubility in water: >1000g/L (23°C). | |
| C1C2C=CC1NC2=O | |
| 109.13 | |
| 109.13 | |
| (±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98% |
| (c=1, CH2Cl2) (1cm cell), (c=1, CH2Cl2) (1cm cell), 2 max. at 400nm | |
| 100.0 | |
| >110°C | |
| 97.5% min. (GC) | |
| C6H7NO | |
| 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one | |
| DDUFYKNOXPZZIW-UHFFFAOYSA-N | |
| 3-azabicyclo[2.2.1]hept-5-en-2-one | |
| 548678 | |
| 98% |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only