missing translation for 'onlineSavingsMsg'
Learn More

2-Anilinopyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3629695
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69201CB 250mg Amber Glass Bottle
CC69201DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC69201CB Supplier Thermo Scientific Chemicals Supplier No. CC69201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 450368-25-9
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD09864415
InChI Key ZPIYMRAJXNMUPE-UHFFFAOYSA-N
Synonym 2-phenylamino pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenylamino, 2-anilino-5-pyrimidinecarboxylic acid, 2-anilinopyrimidine-5-carboxylic acid
PubChem CID 23509722
IUPAC Name 2-anilinopyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=C(NC2=CC=CC=C2)N=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 215.21
Percent Purity 97%
Chemical Name or Material 2-Anilinopyrimidine-5-carboxylic acid