missing translation for 'onlineSavingsMsg'
Learn More

2-Anilinopyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. p-4391171
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69204CB 250mg Amber Glass Bottle
CC69204DA 1g Amber Glass Bottle
CC69204DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC69204CB Supplier Thermo Scientific Chemicals Supplier No. CC69204CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108002-87-5
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
InChI Key QNFHXIZSGIGYOU-UHFFFAOYSA-N
Synonym 2-phenylamino pyrimidine-5-carbaldehyde, 2-anilinopyrimidine-5-carboxaldehyde, 2-phenylamino pyrimidine-5-carboxaldehyde, 2-anilinopyrimidine-5-carbaldehyde
PubChem CID 33589542
IUPAC Name 2-anilinopyrimidine-5-carbaldehyde
SMILES C1=CC=C(C=C1)NC2=NC=C(C=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 2-anilinopyrimidine-5-carbaldehyde
Hazard Category GHS Signal Word: Warning