missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Anilinopyrimidine-5-carbaldehyde, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 199.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-anilinopyrimidine-5-carbaldehyde |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |