missing translation for 'onlineSavingsMsg'
Learn More

(2-anilinopyrimidin-5-yl)methylamine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 1093860-47-9
Molecular Formula C11H12N4
Molecular Weight (g/mol) 200.245
MDL Number MFCD11841074
InChI Key IGMKIKUXMJLMOP-UHFFFAOYSA-N
Synonym 5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
PubChem CID 43811051
IUPAC Name 5-(aminomethyl)-N-phenylpyrimidin-2-amine
SMILES C1=CC=C(C=C1)NC2=NC=C(C=N2)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69213CB
SDS View Documents
Thermo Scientific Chemicals
CC69213CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69213DA
SDS View Documents
Thermo Scientific Chemicals
CC69213DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1093860-47-9
200.245
IGMKIKUXMJLMOP-UHFFFAOYSA-N
43811051
C1=CC=C(C=C1)NC2=NC=C(C=N2)CN
C11H12N4
MFCD11841074
5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
5-(aminomethyl)-N-phenylpyrimidin-2-amine

Specifications

1093860-47-9
100.0
Amber Glass Bottle
MFCD11841074
5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
C1=CC=C(C=C1)NC2=NC=C(C=N2)CN
200.245
200.24
(2-Anilinopyrimidin-5-yl)methylamine
94.5
95%
C11H12N4
250mg
IGMKIKUXMJLMOP-UHFFFAOYSA-N
5-(aminomethyl)-N-phenylpyrimidin-2-amine
43811051
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'