missing translation for 'onlineSavingsMsg'
Learn More

2-Anilinoethanol 98.0+%, TCI America™

Catalog No. p-7116915
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A046625G 25 g
A0466500G 500 g
2 options

Catalog No. A046625G

Supplier: TCI America A046625G

Only null left

Chemical Identifiers

CAS 122-98-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00002832
InChI Key MWGATWIBSKHFMR-UHFFFAOYSA-N
Synonym 2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700
PubChem CID 31235
IUPAC Name 2-anilinoethanol
SMILES C1=CC=C(C=C1)NCCO

Specifications

Melting Point -30°C
Boiling Point 268°C
Quantity 25 g
UN Number 2810
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Anilinoethanol
EINECSNumber (3)-0152&(9)-0054
RTECSNumber KJ7175000
TSCA Yes