missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amylpyridine 98.0+%, TCI America™
$87.60 - $87.60
Chemical Identifiers
| CAS | 2294-76-0 |
|---|---|
| Molecular Formula | C10H15N |
| Molecular Weight (g/mol) | 149.237 |
| MDL Number | MFCD00051828 |
| InChI Key | HSDXVAOHEOSTFZ-UHFFFAOYSA-N |
| Synonym | 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8 |
| PubChem CID | 16800 |
| IUPAC Name | 2-pentylpyridine |
| SMILES | CCCCCC1=CC=CC=N1 |
Chemical Identifiers
| 2294-76-0 | |
| 149.237 | |
| HSDXVAOHEOSTFZ-UHFFFAOYSA-N | |
| 16800 | |
| CCCCCC1=CC=CC=N1 |
| C10H15N | |
| MFCD00051828 | |
| 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8 | |
| 2-pentylpyridine |
Specifications
| 2294-76-0 | |
| 207°C | |
| MFCD00051828 | |
| 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8 | |
| CCCCCC1=CC=CC=N1 | |
| 149.237 | |
| 149.24 | |
| Liquid |
| Yellow | |
| C10H15N | |
| 5 g | |
| HSDXVAOHEOSTFZ-UHFFFAOYSA-N | |
| 2-pentylpyridine | |
| 16800 | |
| ≥98.0% (GC,T) | |
| 2-Amylpyridine |
Safety and Handling
TSCA : Yes