missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-4-carboxamide, 95%

Catalog No. AAL1984303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1984303 1 g
AAL1984306 5 g
AAL1984314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1984303 Supplier Thermo Scientific Chemicals Supplier No. L1984303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13538-42-6 | C6H7N3O | 137.14 g/mol

2-Aminopyridine-4-carboxamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyridine-4-carboxamide is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13538-42-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD03791262
InChI Key AGSDASDGMNDAIE-UHFFFAOYSA-N
PubChem CID 351653
IUPAC Name 2-aminopyridine-4-carboxamide
SMILES NC(=O)C1=CC=NC(N)=C1

Specifications

Melting Point 252°C to 258°C (decomposition)
Quantity 1 g
Beilstein 115924
Synonym 2-aminoisonicotinamide, 2-amino-isonicotinamide, 2-amino-4-pyridinecarboxamide, 4-pyridinecarboxamide, 2-amino, 4-pyridinecarboxamide,2-amino, 2-amino isonicotinamide, pubchem16603, acmc-1bppf, 2-amino-4-pyridinecarbamide, 2-azanylpyridine-4-carboxamide
Solubility Information Slightly soluble in water.
Formula Weight 137.14
Percent Purity 95%
Chemical Name or Material 2-Aminopyridine-4-carboxamide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only