missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopyridine-4-boronic acid pinacol ester, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 161°C to 162°C |
| Color | Colorless to Yellow |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 220.08 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Aminopyridine-4-boronic acid pinacol ester |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only