missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-3-carboxaldehyde, 98%

Catalog No. AAL1962003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962003 1 g
AAL1962006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1962003 Supplier Thermo Scientific Chemicals Supplier No. L1962003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7521-41-7 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7521-41-7
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD01830382
InChI Key NXMFJCRMSDRXLD-UHFFFAOYSA-N
Synonym 2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
PubChem CID 737633
IUPAC Name 2-aminopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)N)C=O

Specifications

Melting Point 99°C to 102°C
Quantity 1 g
Beilstein 109598
Sensitivity Air sensitive
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material 2-Aminopyridine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only