missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopyrazine 98.0+%, TCI America™
Supplier: TCI America A098925G
Specifications
| 2-Aminopyrazine | |
| 121°C | |
| MFCD00006137 | |
| 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine | |
| C1=CN=C(C=N1)N | |
| 95.105 | |
| 95.11 | |
| Crystalline Powder |
| 5049-61-6 | |
| C4H5N3 | |
| 25 g | |
| XFTQRUTUGRCSGO-UHFFFAOYSA-N | |
| pyrazin-2-amine | |
| 78747 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 5049-61-6 | |
| 95.105 | |
| XFTQRUTUGRCSGO-UHFFFAOYSA-N | |
| 78747 | |
| C1=CN=C(C=N1)N |
| C4H5N3 | |
| MFCD00006137 | |
| 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine | |
| pyrazin-2-amine |
Safety and Handling
TSCA : No