missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine 98.0+%, TCI America™

Catalog No. A1111200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1111200MG 200 mg
A11111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1111200MG Supplier TCI America Supplier No. A1111200MG

Chemical Identifiers

CAS 452-06-2
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005566
InChI Key MWBWWFOAEOYUST-UHFFFAOYSA-N
Synonym 2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine
PubChem CID 9955
ChEBI CHEBI:479072
IUPAC Name 7H-purin-2-amine
SMILES NC1=NC=C2NC=NC2=N1

Specifications

Melting Point 278°C
Color Yellow
Quantity 200 mg
Formula Weight 135.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopurine
RTECSNumber UO7475000
TSCA No