missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine, 98%

Catalog No. AAJ64919MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64919MD 250 mg
AAJ64919ME 500 mg
AAJ6491903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ64919MD Supplier Thermo Scientific Chemicals Supplier No. J64919MD

A protein kinase R inhibitor | CAS: 452-06-2 | C5H5N5 | 135.13 g/mol

A protein kinase R inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-06-2
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005566
InChI Key MWBWWFOAEOYUST-UHFFFAOYSA-N
Synonym 2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine
PubChem CID 9955
ChEBI CHEBI:479072
IUPAC Name 7H-purin-2-amine
SMILES NC1=NC=C2NC=NC2=N1

Specifications

Melting Point 280°C to 282°C
Color White to Yellow
Odor Odorless
Quantity 250 mg
Formula Weight 135.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Aminopurine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 207-197-4
RTECSNumber UO7475000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only