missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenylboronic acid hydrochloride, 97%

CAS: 863753-30-4 | C6H9BClNO2 | 173.40 g/mol

$788.54 - $788.54

Chemical Identifiers

CAS 863753-30-4
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD02258096
InChI Key WPDASZCYRKGSTO-UHFFFAOYSA-N
Synonym 2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl
PubChem CID 16427090
SMILES Cl.NC1=CC=CC=C1B(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445310050
SDS View Documents
Thermo Scientific Chemicals
445310050
5 g
Each for $788.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

863753-30-4
173.40
WPDASZCYRKGSTO-UHFFFAOYSA-N
16427090
C6H9BClNO2
MFCD02258096
2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl
Cl.NC1=CC=CC=C1B(O)O

Specifications

863753-30-4
Glass Bottle
MFCD02258096
2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl
Cl.NC1=CC=CC=C1B(O)O
173.40
173.41
2-Aminophenylboronic acid hydrochloride
96% min. (HPLC)
C6H9BClNO2
5 g
WPDASZCYRKGSTO-UHFFFAOYSA-N
(2-aminophenyl)boronic acid;hydrochloride
16427090
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only