missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenylarsonic Acid 98.0+%, TCI America™

Catalog No. A05295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05295G 5 g
A052925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05295G Supplier TCI America Supplier No. A05295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2045-00-3
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
MDL Number MFCD00007630
InChI Key LQCOCUQCZYAYQK-UHFFFAOYSA-N
Synonym o-arsanilic acid, 2-aminobenzenearsonic acid, o-aminobenzenearsonic acid, 2-aminophenyl arsonic acid, arsenical, unii-4p7t2ph7dl, 2-arsanilic acid, o-aminophenylarsonic acid, 4p7t2ph7dl, arsonic acid,as-2-aminophenyl
PubChem CID 73161
IUPAC Name (2-aminophenyl)arsonic acid
SMILES C1=CC=C(C(=C1)N)[As](=O)(O)O

Specifications

Melting Point 153°C
Quantity 5 g
UN Number 3465
Formula Weight 217.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenylarsonic Acid