missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoperimidine Hydrobromide 98.0+%, TCI America™

Catalog No. p-7118266
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51335G 5 g
A513325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A51335G Supplier TCI America Supplier No. A51335G
Only null left

Chemical Identifiers

CAS 40835-96-9
Molecular Formula C11H10BrN3
Molecular Weight (g/mol) 264.126
MDL Number MFCD00012749
InChI Key HFZUTZWRTHBWPV-UHFFFAOYSA-N
PubChem CID 2724235
IUPAC Name 1H-perimidin-2-amine;hydrobromide
SMILES C1=CC2=C3C(=C1)NC(=NC3=CC=C2)N.Br

Specifications

Melting Point 300°C
Quantity 5 g
Formula Weight 264.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoperimidine Hydrobromide [Precipitation reagent for SO4]