missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopentane 97.0+%, TCI America™
Supplier: TCI America A110225ML
Specifications
| 2-Aminopentane | |
| Yellow | |
| C5H13N | |
| 25 mL | |
| 2-Pentylamine | |
| CCCC(C)N | |
| 87.166 | |
| 87.17 | |
| Liquid |
| 63493-28-7 | |
| 91°C | |
| MFCD00008098 | |
| 1106 | |
| IGEIPFLJVCPEKU-UHFFFAOYSA-N | |
| pentan-2-amine | |
| 12246 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 63493-28-7 | |
| 87.166 | |
| IGEIPFLJVCPEKU-UHFFFAOYSA-N | |
| 12246 | |
| CCCC(C)N |
| C5H13N | |
| MFCD00008098 | |
| 2-Pentylamine | |
| pentan-2-amine |
Safety and Handling
EINECSNumber : (2)-0133
TSCA : Yes