missing translation for 'onlineSavingsMsg'
Learn More

2-Aminooxazole 98.0+%, TCI America™

Catalog No. p-7118126
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A28841G 1 g
A28845G 5 g
2 options

Catalog No. A28841G

Supplier: TCI America A28841G

Only null left

Chemical Identifiers

CAS 4570-45-0
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD07364485
InChI Key ACTKAGSPIFDCMF-UHFFFAOYSA-N
Synonym oxazol-2-amine, 2-aminooxazole, oxazole-2-amine, 2-amino-oxazole, 2-oxazolamine, 2-amino-1,3-oxazole, oxazol-2-ylamine, 2-aminoxazole, 1,3-oxazol-2-ylamine, 1,3-oxazole-2-ylamine
PubChem CID 558521
IUPAC Name 1,3-oxazol-2-amine
SMILES NC1=NC=CO1

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 84.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminooxazole
TSCA No
Recommended Storage Refrigerator