missing translation for 'onlineSavingsMsg'
Learn More

2-Aminooxazole, 97%

Catalog No. AAH3436003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3436003 1 g
AAH3436006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3436003 Supplier Thermo Scientific Chemicals Supplier No. H3436003

CAS: 4570-45-0 | C3H4N2O | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4570-45-0
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD07364485
InChI Key ACTKAGSPIFDCMF-UHFFFAOYSA-N
Synonym oxazol-2-amine, 2-aminooxazole, oxazole-2-amine, 2-amino-oxazole, 2-oxazolamine, 2-amino-1,3-oxazole, oxazol-2-ylamine, 2-aminoxazole, 1,3-oxazol-2-ylamine, 1,3-oxazole-2-ylamine
PubChem CID 558521
IUPAC Name 1,3-oxazol-2-amine
SMILES NC1=NC=CO1

Specifications

Melting Point 93°C to 95°C
Quantity 1 g
Formula Weight 84.1
Percent Purity 97%
Chemical Name or Material 2-Aminooxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only