missing translation for 'onlineSavingsMsg'
Learn More

2-Aminooctane 98.0+%, TCI America™

Catalog No. M03371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03371G Supplier TCI America Supplier No. M03371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 693-16-3
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008103
InChI Key HBXNJMZWGSCKPW-UHFFFAOYSA-N
Synonym 2-aminooctane, 2-octanamine, 2-octylamine, 1-methylheptylamine, heptylamine, 1-methyl, 2-caprylamine, sec-octylamine, 1-methyl heptylamine, oct-2-ylamine, sec-octanamine
PubChem CID 12735
IUPAC Name octan-2-amine
SMILES CCCCCCC(C)N

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
UN Number 2734
Formula Weight 129.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminooctane
EINECSNumber (2)-0133
RTECSNumber MK1210000
TSCA Yes