Learn More
2-Aminonicotinamide, 97%
CAS: 13438-65-8 | C6H7N3O | 137.142 g/mol
$109.97 - $418.38
Chemical Identifiers
| CAS | 13438-65-8 |
|---|---|
| Molecular Formula | C6H7N3O |
| Molecular Weight (g/mol) | 137.142 |
| MDL Number | MFCD00128873 |
| InChI Key | HTPCDVLWYUXWQR-UHFFFAOYSA-N |
| Synonym | 2-aminonicotinamide, 2-amino-nicotinamide, 2-amino nicotinamide, 3-pyridinecarboxamide,2-amino, 3-pyridinecarboxamide, ar-amino, 2-amino-3-pyridinecarboxamide, amino-nicotinamide, ar-aminonicotinamide, ar-amino-3-pyridinecarboxamide, nicotinamide, amino |
| PubChem CID | 506623 |
| IUPAC Name | 2-aminopyridine-3-carboxamide |
| SMILES | C1=CC(=C(N=C1)N)C(=O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6620203
|
Thermo Scientific Chemicals
H6620203 |
1 g |
Each for $109.97
|
|
|||||
|
AAH6620206
|
Thermo Scientific Chemicals
H6620206 |
5 g |
Each for $418.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13438-65-8 | |
| 137.142 | |
| HTPCDVLWYUXWQR-UHFFFAOYSA-N | |
| 506623 | |
| C1=CC(=C(N=C1)N)C(=O)N |
| C6H7N3O | |
| MFCD00128873 | |
| 2-aminonicotinamide, 2-amino-nicotinamide, 2-amino nicotinamide, 3-pyridinecarboxamide,2-amino, 3-pyridinecarboxamide, ar-amino, 2-amino-3-pyridinecarboxamide, amino-nicotinamide, ar-aminonicotinamide, ar-amino-3-pyridinecarboxamide, nicotinamide, amino | |
| 2-aminopyridine-3-carboxamide |
Specifications
| 13438-65-8 | |
| Yellow | |
| MFCD00128873 | |
| UN2811 | |
| 2-aminonicotinamide, 2-amino-nicotinamide, 2-amino nicotinamide, 3-pyridinecarboxamide,2-amino, 3-pyridinecarboxamide, ar-amino, 2-amino-3-pyridinecarboxamide, amino-nicotinamide, ar-aminonicotinamide, ar-amino-3-pyridinecarboxamide, nicotinamide, amino | |
| C1=CC(=C(N=C1)N)C(=O)N | |
| 137.142 | |
| 137.14 | |
| Solid |
| 194°C to 200°C | |
| C6H7N3O | |
| 1 g | |
| 3613 | |
| HTPCDVLWYUXWQR-UHFFFAOYSA-N | |
| 2-aminopyridine-3-carboxamide | |
| 506623 | |
| 97% | |
| 2-Aminonicotinamide |
Safety and Handling
GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
H302-H317-H319
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only