missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinaldehyde 97.0+%, TCI America™

Catalog No. A18181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18181G 1 g
A18185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18181G Supplier TCI America Supplier No. A18181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7521-41-7
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
InChI Key NXMFJCRMSDRXLD-UHFFFAOYSA-N
Synonym 2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
PubChem CID 737633
IUPAC Name 2-aminopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)N)C=O

Specifications

Melting Point 98°C
Quantity 1 g
Formula Weight 122.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminonicotinaldehyde
TSCA No
Recommended Storage Refrigerator