missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)pyridine, 99%

Catalog No. AC104070250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104070250 25 mL Glass bottle
AC104071000 100 mL Glass bottle
AC104075000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104070250 Supplier Thermo Scientific Chemicals Supplier No. 104070250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3731-51-9 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3731-51-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006360
InChI Key WOXFMYVTSLAQMO-UHFFFAOYSA-N
Synonym 2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
PubChem CID 19509
ChEBI CHEBI:81387
IUPAC Name pyridin-2-ylmethanamine
SMILES NCC1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Colorless to Yellow or Orange
Density 1.0400g/mL
Boiling Point 82.0°C to 85.0°C (12.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5420 to 1.5460
Quantity 25 mL
Specific Gravity 1.04
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-090-5

RUO – Research Use Only