missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)piperidine 99.0+%, TCI America™

Catalog No. A25175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25175G Supplier TCI America Supplier No. A25175G

Chemical Identifiers

CAS 22990-77-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00129011
InChI Key RHPBLLCTOLJFPH-UHFFFAOYNA-N
Synonym 2-piperidylmethylamine, 2-aminomethyl piperidine, 2-piperidinemethanamine, 2-aminomethylpiperidine, 2-aminomethyl-1-piperidine, 1-piperidin-2-yl methanamine, piperidine-2-methylamine, piperidine, 2-aminomethyl, pubchem6725, 2-aminomethyl-piperdine
PubChem CID 90865
IUPAC Name 1-(piperidin-2-yl)methanamine
SMILES NCC1CCCCN1

Specifications

Color Colorless
Boiling Point 67°C
Quantity 5 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)piperidine
EINECSNumber (5)-5685
TSCA No