missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)piperidine, 98%

Catalog No. p-7048029
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2012206 5 g
AAL2012214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2012206 Supplier Thermo Scientific Chemicals Supplier No. L2012206

CAS: 22990-77-8 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22990-77-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00129011
InChI Key RHPBLLCTOLJFPH-UHFFFAOYNA-N
Synonym 2-piperidylmethylamine, 2-aminomethyl piperidine, 2-piperidinemethanamine, 2-aminomethylpiperidine, 2-aminomethyl-1-piperidine, 1-piperidin-2-yl methanamine, piperidine-2-methylamine, piperidine, 2-aminomethyl, pubchem6725, 2-aminomethyl-piperdine
PubChem CID 90865
IUPAC Name piperidin-2-ylmethanamine
SMILES NCC1CCCCN1

Specifications

Density 0.942
Boiling Point 63°C to 66°C (12 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.485
Quantity 5 g
UN Number UN2735
Beilstein 102994
Sensitivity Air Sensitive
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 2-(Aminomethyl)piperidine
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 245-371-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only