missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)indole, 97%

Catalog No. p-7036040
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54281MD 250 mg
AAH5428103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54281MD Supplier Thermo Scientific Chemicals Supplier No. H54281MD

CAS: 21109-25-1 | C9H10N2 | 146.193 g/mol

2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 21109-25-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD03422512
InChI Key RNAODKZCUVVPEN-UHFFFAOYSA-N
Synonym 1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
PubChem CID 582331
IUPAC Name 1H-indol-2-ylmethanamine
SMILES C1=CC=C2C(=C1)C=C(N2)CN

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 2-(Aminomethyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only