missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Aminomethyl)-5-methylpyrazine 98.0+%, TCI America™
Supplier: TCI America A11545G
Specifications
| 2-(Aminomethyl)-5-methylpyrazine | |
| Yellow | |
| C6H9N3 | |
| 5 g | |
| MPBCUCGKHDEUDD-UHFFFAOYSA-N | |
| (5-methylpyrazin-2-yl)methanamine | |
| 2756496 | |
| ≥98.0% (GC,T) |
| 132664-85-8 | |
| 94°C | |
| MFCD01314542 | |
| 2735 | |
| CC1=CN=C(C=N1)CN | |
| 123.159 | |
| 123.16 | |
| Liquid |
Chemical Identifiers
| 132664-85-8 | |
| 123.159 | |
| MPBCUCGKHDEUDD-UHFFFAOYSA-N | |
| (5-methylpyrazin-2-yl)methanamine |
| C6H9N3 | |
| MFCD01314542 | |
| 2756496 | |
| CC1=CN=C(C=N1)CN |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator