missing translation for 'onlineSavingsMsg'
Learn More

2-Aminomethyl-1-Boc-piperidine, 95%

Catalog No. p-7036246
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5450703 1 g
AAH5450706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5450703 Supplier Thermo Scientific Chemicals Supplier No. H5450703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 370069-31-1 | C11H22N2O2 | 214.309 g/mol

Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.

Solubility
Soluble in DMSO.

Notes
Refrigerated (2-8°C). Incompatible with oxidizing agents.

Chemical Identifiers

CAS 370069-31-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01317791
InChI Key PTVRCUVHYMGECC-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-boc-piperidine, tert-butyl 2-aminomethyl piperidine-1-carboxylate, 2-aminomethyl-1-n-boc-piperidine, 1-boc-2-aminomethyl-piperidine, 1-boc-2-aminomethyl piperidine, 1-boc-2-aminomethylpiperidine, 2-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-2-aminomethyl piperidine
PubChem CID 2756482
IUPAC Name tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1CN

Specifications

Density 1.012
Flash Point >110°C (230°F)
Refractive Index 1.4745
Quantity 1 g
UN Number UN3082
Solubility Information Soluble in DMSO.
Formula Weight 214.31
Percent Purity 95%
Chemical Name or Material 2-Aminomethyl-1-Boc-piperidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only