missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoisonicotinic Acid 97.0+%, TCI America™
$212.51 - $775.44
Chemical Identifiers
| CAS | 13362-28-2 |
|---|---|
| Molecular Formula | C6H6N2O2 |
| Molecular Weight (g/mol) | 138.126 |
| MDL Number | MFCD00233714 |
| InChI Key | QMKZZQPPJRWDED-UHFFFAOYSA-N |
| Synonym | 2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j |
| PubChem CID | 278396 |
| IUPAC Name | 2-aminopyridine-4-carboxylic acid |
| SMILES | C1=CN=C(C=C1C(=O)O)N |
Chemical Identifiers
| 13362-28-2 | |
| 138.126 | |
| QMKZZQPPJRWDED-UHFFFAOYSA-N | |
| 278396 | |
| C1=CN=C(C=C1C(=O)O)N |
| C6H6N2O2 | |
| MFCD00233714 | |
| 2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j | |
| 2-aminopyridine-4-carboxylic acid |
Specifications
| 13362-28-2 | |
| Yellow | |
| MFCD00233714 | |
| 2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j | |
| C1=CN=C(C=C1C(=O)O)N | |
| 138.126 | |
| 138.13 | |
| Crystalline Powder |
| 290°C | |
| C6H6N2O2 | |
| 1 g | |
| QMKZZQPPJRWDED-UHFFFAOYSA-N | |
| 2-aminopyridine-4-carboxylic acid | |
| 278396 | |
| ≥97.0% (HPLC,T) | |
| 2-Aminoisonicotinic Acid |
Safety and Handling
TSCA : No