missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoisonicotinamide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 249°C |
| Quantity | 5 g |
| UN Number | 2811 |
| Formula Weight | 137.14 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Aminoisonicotinamide |
Safety and Handling
| TSCA | No |