missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisonicotinamide 98.0+%, TCI America™

Catalog No. A28991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28991G 1 g
A28995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28991G Supplier TCI America Supplier No. A28991G

Chemical Identifiers

CAS 13538-42-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD03791262
InChI Key AGSDASDGMNDAIE-UHFFFAOYSA-N
Synonym 2-aminoisonicotinamide, 2-amino-isonicotinamide, 2-amino-4-pyridinecarboxamide, 4-pyridinecarboxamide, 2-amino, 4-pyridinecarboxamide,2-amino, 2-amino isonicotinamide, pubchem16603, acmc-1bppf, 2-amino-4-pyridinecarbamide, 2-azanylpyridine-4-carboxamide
PubChem CID 351653
IUPAC Name 2-aminopyridine-4-carboxamide
SMILES NC(=O)C1=CC=NC(N)=C1

Specifications

Melting Point 249°C
Quantity 1 g
UN Number 2811
Formula Weight 137.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoisonicotinamide