missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid methyl ester hydrochloride, 99%

Catalog No. p-7029116
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2655003 1 g
AAH2655006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2655003 Supplier Thermo Scientific Chemicals Supplier No. H2655003

CAS: 15028-41-8 | C5H12ClNO2 | 153.606 g/mol

Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 15028-41-8
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00214247
InChI Key NVWZNEDLYYLQJC-UHFFFAOYSA-N
Synonym methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
PubChem CID 13258034
IUPAC Name methyl 2-amino-2-methylpropanoate;hydrochloride
SMILES CC(C)(C(=O)OC)N.Cl

Specifications

Melting Point ∽185°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid methyl ester hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only